Assay Details
Target Gene Details
Entrez Gene ID: | 5154 |
Gene Name: | platelet derived growth factor subunit A |
Gene Aliases: |
PDGF-A, PDGF1 |
Location: |
Chr.7:497245-520668 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFA | NM_002607.5 | 4 | 1254 | NP_002598.4 |
| NM_033023.4 | 4 | 1254 | NP_148983.1 | |
| XM_011515415.1 | 5 | 746 | XP_011513717.1 | |
| XM_011515416.1 | 5 | 750 | XP_011513718.1 | |
| XM_011515417.1 | 5 | 746 | XP_011513719.1 | |
| XM_011515418.1 | 5 | 750 | XP_011513720.1 | |
| XM_011515419.2 | 4 | 454 | XP_011513721.1 | |
| XM_017012289.1 | 4 | 487 | XP_016867778.1 | |
| A09204.1 | 4 | 447 | CAA00830.1 | |
| AB451309.1 | 4 | 411 | ||
| AB451439.1 | 4 | 411 | ||
| AK292217.1 | 4 | 625 | ||
| AU118732.1 | 4 | 582 | ||
| BC109246.1 | 2 | 1274 | AAI09247.1 | |
| BE336646.1 | 3 | 368 | ||
| X03795.1 | 4 | 798 | CAA27421.1 | |
| X06374.1 | 4 | 814 | CAA29677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950782 | Chr.7:408935 - 587963 on Build GRCh38 | Deletion |
|
| nsv1119098 | Chr.7:510263 - 510963 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map