Assay Details
Target Gene Details
Entrez Gene ID: | 127665 |
Gene Name: | zinc finger protein 648 |
Gene Aliases: |
- |
Location: |
Chr.1:182053740-182205330 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF648 | NM_001009992.1 | 2 | 597 | NP_001009992.1 |
| XM_017000301.1 | 4 | 4024 | XP_016855790.1 | |
| XM_017000302.1 | 6 | 4530 | XP_016855791.1 | |
| AK128654.1 | 2 | 597 | ||
| BC137223.1 | 2 | 500 | ||
| BC137224.1 | 2 | 500 |
More Information
Set Membership: |
|

Genomic Map