Assay Details
Target Gene Details
Entrez Gene ID: | 10368 |
Gene Name: | calcium voltage-gated channel auxiliary subunit gamma 3 |
Gene Aliases: |
- |
Location: |
Chr.16:24255553-24362416 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNG3 | NM_006539.3 | 1 | 686 | NP_006530.1 |
| AK055453.1 | 1 | 85 | ||
| AK095553.1 | 1 | 684 | ||
| BC037899.1 | 1 | 691 | AAH37899.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833174 | Chr.16:24077853 - 24258848 on Build GRCh38 | Gain |
|
| esv34056 | Chr.16:24123791 - 24318980 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map