Assay Details
Target Gene Details
Entrez Gene ID: | 81697 |
Gene Name: | olfactory receptor family 2 subfamily B member 2 |
Gene Aliases: |
OR2B2Q, OR2B9, OR6-1, dJ193B12.4, hs6M1-10 |
Location: |
Chr.6:27911185-27912396 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR2B2 | NM_033057.2 | 1 | 96 | NP_149046.2 |
| BC136881.1 | 1 | 96 | ||
| BC136901.1 | 1 | 96 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830619 | Chr.6:27904524 - 28094766 on Build GRCh38 | Loss |
|
| nsv601208 | Chr.6:27842848 - 28001226 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map