Assay Details
Target Gene Details
Entrez Gene ID: | 55258 |
Gene Name: | threonine synthase like 2 |
Gene Aliases: |
SOFAT, THS2, TSH2 |
Location: |
Chr.2:88170295-88186637 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THNSL2 | NM_001244676.1 | NP_001231605.1 | ||
| NM_001244678.1 | NP_001231607.1 | |||
| NM_018271.4 | NP_060741.3 | |||
| XM_005264400.4 | XP_005264457.1 | |||
| XM_005264401.4 | XP_005264458.1 | |||
| XM_005264403.4 | XP_005264460.1 | |||
| XM_005264405.4 | XP_005264462.1 | |||
| XM_006712041.3 | XP_006712104.1 | |||
| XM_006712044.3 | XP_006712107.1 | |||
| XM_011532954.2 | XP_011531256.1 | |||
| XM_011532955.2 | XP_011531257.1 | |||
| AI313402.1 | ||||
| AK001778.1 | BAA91904.1 | |||
| AK092823.1 | ||||
| AK095303.1 | ||||
| AL832746.1 | ||||
| BC035315.1 | AAH35315.1 | |||
| BC047758.2 | AAH47758.2 | |||
| BC064423.1 | AAH64423.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv61n16 | Chr.2:87343737 - 90254660 on Build GRCh38 | Deletion |
|
| dgv6947n54 | Chr.2:87815738 - 88850700 on Build GRCh38 | Gain |
|
| nsv1072005 | Chr.2:87360367 - 90256528 on Build GRCh38 | Deletion |
|
| nsv1148611 | Chr.2:87360364 - 90256525 on Build GRCh38 | Deletion |
|
| nsv834289 | Chr.2:88083076 - 88251327 on Build GRCh38 | Gain |
|
| nsv582384 | Chr.2:87987149 - 88773214 on Build GRCh38 | Loss |
|
| nsv582386 | Chr.2:88035188 - 89208087 on Build GRCh38 | Loss |
|
| nsv582383 | Chr.2:87915816 - 88527080 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map