Assay Details
Target Gene Details
Entrez Gene ID: | 8907 |
Gene Name: | adaptor related protein complex 1 mu 1 subunit |
Gene Aliases: |
AP47, CLAPM2, CLTNM, MU-1A |
Location: |
Chr.19:16197854-16235345 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1M1 | NM_001130524.1 | NP_001123996.1 | ||
| NM_032493.3 | NP_115882.1 | |||
| XM_017027420.1 | XP_016882909.1 | |||
| XM_017027421.1 | XP_016882910.1 | |||
| AB209808.1 | BAD93045.1 | |||
| AF290613.1 | AAK28024.1 | |||
| AK027528.1 | ||||
| AK293188.1 | ||||
| AK297824.1 | ||||
| BC017469.2 | AAH17469.1 | |||
| DQ059565.1 | AAY54246.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833767 | Chr.19:16121391 - 16347760 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map