Assay Details
Target Gene Details
Entrez Gene ID: | 3553 |
Gene Name: | interleukin 1 beta |
Gene Aliases: |
IL-1, IL1-BETA, IL1F2 |
Location: |
Chr.2:112829758-112836842 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1B | NM_000576.2 | 5 | 467 | NP_000567.1 |
| XM_017003988.1 | 4 | 468 | XP_016859477.1 | |
| AB451494.1 | ||||
| AF043335.1 | AAC03536.1 | |||
| AK313983.1 | 4 | 1187 | ||
| BC008678.1 | 5 | 467 | AAH08678.1 | |
| BT007213.1 | 4 | 380 | AAP35877.1 | |
| CR407679.1 | 4 | 380 | CAG28607.1 | |
| K02770.1 | 5 | 466 | AAA36106.1 | |
| M15330.1 | 5 | 466 | AAA59135.1 | |
| M54933.1 | 5 | 467 | AAA59136.1 | |
| X02532.1 | 5 | 457 | CAA26372.1 | |
| X56087.1 | 5 | 437 | CAA39567.1 |
More Information
Set Membership: |
|

Genomic Map