Assay Details
Target Gene Details
Entrez Gene ID: | 27333 |
Gene Name: | golgi integral membrane protein 4 |
Gene Aliases: |
GIMPC, GOLPH4, GPP130, P138 |
Location: |
Chr.3:168008672-168095925 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLIM4 | NM_001308155.1 | 11 | 2127 | NP_001295084.1 |
| NM_014498.4 | 12 | 2211 | NP_055313.1 | |
| XM_005247364.2 | 12 | 2213 | XP_005247421.1 | |
| XM_005247365.2 | 11 | 2129 | XP_005247422.1 | |
| XM_017006183.1 | 11 | 1609 | XP_016861672.1 | |
| AK307557.1 | 12 | 1916 | ||
| AK307718.1 | 12 | 1886 | ||
| U55853.1 | 12 | 1584 | AAB58419.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829787 | Chr.3:168024030 - 168222588 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map