Assay Details
Target Gene Details
Entrez Gene ID: | 11120 |
Gene Name: | butyrophilin subfamily 2 member A1 |
Gene Aliases: |
BK14H9.1, BT2.1, BTF1, BTN2.1, DJ3E1.1 |
Location: |
Chr.6:26457904-26476621 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTN2A1 | NM_001197233.2 | 7 | 2461 | NP_001184162.1 |
| NM_001197234.2 | NP_001184163.1 | |||
| NM_007049.4 | 8 | 2573 | NP_008980.1 | |
| NM_078476.3 | 8 | 2802 | NP_510961.1 | |
| AB209777.1 | BAD93014.1 | |||
| AK093886.1 | 1 | 2001 | ||
| AW007668.1 | 1 | 382 | ||
| BC016661.1 | 8 | 2745 | AAH16661.1 | |
| BX452202.2 | 1 | 454 | ||
| BX538050.1 | 7 | 2116 | CAD97989.1 | |
| U90142.1 | 11 | 2348 | AAC02650.1 | |
| U90543.1 | 8 | 2514 | AAB53421.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023453 | Chr.6:25853602 - 26528022 on Build GRCh38 | Gain |
|
| dgv5928n100 | Chr.6:26428086 - 26471368 on Build GRCh38 | Loss |
|
| nsv1031938 | Chr.6:26466954 - 26501669 on Build GRCh38 | Loss |
|
| nsv965689 | Chr.6:26456805 - 26469259 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map