Assay Details
Target Gene Details
Entrez Gene ID: | 51449 |
Gene Name: | prenylcysteine oxidase 1 |
Gene Aliases: |
PCL1 |
Location: |
Chr.2:70258099-70281185 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCYOX1 | NM_016297.3 | NP_057381.3 | ||
| AB020715.1 | ||||
| AF181490.1 | AAF16937.1 | |||
| AK225302.1 | ||||
| AK296478.1 | ||||
| AK303043.1 | ||||
| AK314453.1 | ||||
| AK316013.1 | ||||
| AY359063.1 | AAQ89422.1 | |||
| BC007029.1 | AAH07029.1 | |||
| BC033815.1 | AAH33815.1 | |||
| BC051891.1 | AAH51891.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518294 | Chr.2:70111212 - 70301469 on Build GRCh38 | Loss |
|
| esv2760537 | Chr.2:70270692 - 70413622 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map