Assay Details
Target Gene Details
Entrez Gene ID: | 4306 |
Gene Name: | nuclear receptor subfamily 3 group C member 2 |
Gene Aliases: |
MCR, MLR, MR, NR3C2VIT |
Location: |
Chr.4:148078764-148445287 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR3C2 | NM_000901.4 | 9 | 4441 | NP_000892.2 |
| NM_001166104.1 | 8 | 4090 | NP_001159576.1 | |
| XM_011531975.1 | 9 | 4453 | XP_011530277.1 | |
| XM_011531976.2 | 9 | 4750 | XP_011530278.1 | |
| XM_011531977.2 | 9 | 4180 | XP_011530279.1 | |
| XM_017008217.1 | 9 | 4738 | XP_016863706.1 | |
| BC111758.1 | AAI11759.1 | |||
| M16801.1 | 9 | 4294 | AAA59571.1 |
More Information
Set Membership: |
|

Genomic Map