Assay Details
Target Gene Details
Entrez Gene ID: | 27288 |
Gene Name: | RNA binding motif protein, X-linked like 2 |
Gene Aliases: |
HNRNPG-T, HNRNPGT, HNRPGT |
Location: |
Chr.11:7088934-7091148 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBMXL2 | NM_014469.4 | NP_055284.3 | ||
| BX091666.1 | 1 | 453 |
More Information
Set Membership: |
|

Genomic Map