Assay Details
Target Gene Details
Entrez Gene ID: | 81565 |
Gene Name: | nudE neurodevelopment protein 1 like 1 |
Gene Aliases: |
EOPA, MITAP1, NDE1L1, NDE2, NUDEL |
Location: |
Chr.17:8435852-8472744 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDEL1 | NM_001025579.2 | NP_001020750.1 | ||
| NM_030808.4 | NP_110435.1 | |||
| XM_011524013.1 | XP_011522315.1 | |||
| XM_017025183.1 | XP_016880672.1 | |||
| XM_017025184.1 | XP_016880673.1 | |||
| XM_017025185.1 | XP_016880674.1 | |||
| XM_017025186.1 | XP_016880675.1 | |||
| XM_017025187.1 | XP_016880676.1 | |||
| XM_017025188.1 | XP_016880677.1 | |||
| AF182078.1 | AAG43425.1 | |||
| AF217798.3 | AAF24516.3 | |||
| AK056014.1 | ||||
| AK299559.1 | ||||
| AL832648.1 | CAD89957.3 | |||
| AY004871.1 | AAF97497.1 | |||
| BC026101.2 | AAH26101.1 | |||
| BP201698.1 | ||||
| CX166766.1 | ||||
| DB176966.1 | ||||
| DN993952.1 |
More Information
Set Membership: |
|

Genomic Map