Assay Details
Target Gene Details
Entrez Gene ID: | 10485 |
Gene Name: | chromosome 1 open reading frame 61 |
Gene Aliases: |
CROC4 |
Location: |
Chr.1:156404252-156456649 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 10485 |
Gene Name: | chromosome 1 open reading frame 61 |
Gene Aliases: |
CROC4 |
Location: |
Chr.1:156404252-156456649 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |