Assay Details
Target Gene Details
Entrez Gene ID: | 8824 |
Gene Name: | carboxylesterase 2 |
Gene Aliases: |
CE-2, CES2A1, PCE-2, iCE |
Location: |
Chr.16:66934444-66945096 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CES2 | NM_003869.5 | NP_003860.2 | ||
| NM_198061.2 | NP_932327.1 | |||
| NR_036684.1 | ||||
| XM_011523421.2 | XP_011521723.1 | |||
| XM_017023818.1 | XP_016879307.1 | |||
| AK095522.1 | ||||
| AK222724.1 | BAD96444.1 | |||
| AK290482.1 | ||||
| AL713761.1 | CAD28531.1 | |||
| BC032095.2 | AAH32095.1 | |||
| BC071874.1 | AAH71874.1 | |||
| BC098417.1 | AAH98417.1 | |||
| BX538086.1 | CAD98009.1 | |||
| D50579.1 | BAA23606.1 | |||
| U60553.1 | AAB03611.1 | |||
| Y09616.1 | CAA70831.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827706 | Chr.16:66894354 - 67009614 on Build GRCh38 | Loss |
|
| dgv526e214 | Chr.16:66940449 - 67031371 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map