Assay Details
Target Gene Details
Entrez Gene ID: | 5118 |
Gene Name: | procollagen C-endopeptidase enhancer |
Gene Aliases: |
PCPE, PCPE-1, PCPE1 |
Location: |
Chr.7:100602259-100608175 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCOLCE | NM_002593.3 | NP_002584.2 | ||
| AB008549.1 | BAA23281.1 | |||
| AK313748.1 | ||||
| BC000574.2 | AAH00574.1 | |||
| BC033205.1 | AAH33205.1 | |||
| L33799.1 | AAA61949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
| nsv951386 | Chr.7:100566478 - 100682977 on Build GRCh38 | Deletion |
|
| nsv831072 | Chr.7:100564880 - 100765212 on Build GRCh38 | Loss |
|
| nsv5868 | Chr.7:100572375 - 100617023 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map