Assay Details
Target Gene Details
Entrez Gene ID: | 23116 |
Gene Name: | family with sequence similarity 179 member B |
Gene Aliases: |
KIAA0423 |
Location: |
Chr.14:44962190-45074431 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM179B | NM_001308120.1 | 1 | 1017 | NP_001295049.1 |
| NM_015091.3 | 1 | 1017 | NP_055906.2 | |
| NR_131765.1 | 1 | 1017 | ||
| XM_011536571.1 | 1 | 988 | XP_011534873.1 | |
| XM_017021098.1 | 1 | 988 | XP_016876587.1 | |
| XM_017021099.1 | 1 | 988 | XP_016876588.1 | |
| XM_017021100.1 | 1 | 988 | XP_016876589.1 | |
| AB007883.2 | 1 | 999 | ||
| AK096934.1 | 1 | 988 | ||
| AK295185.1 | 1 | 1017 | ||
| AK307722.1 | 2 | 814 | ||
| AK311034.1 | 1 | 988 | ||
| BC057255.2 | 1 | 994 | AAH57255.1 | |
| BX648723.1 | 1 | 968 | ||
| CR749557.1 | 1 | 995 | CAH18354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1904n100 | Chr.14:44932303 - 45028704 on Build GRCh38 | Loss |
|
| nsv1259 | Chr.14:44950120 - 44975457 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map