Assay Details
Target Gene Details
Entrez Gene ID: | 993 |
Gene Name: | cell division cycle 25A |
Gene Aliases: |
CDC25A2 |
Location: |
Chr.3:48157178-48188658 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC25A | NM_001789.2 | NP_001780.2 | ||
| NM_201567.1 | NP_963861.1 | |||
| XM_006713434.2 | XP_006713497.1 | |||
| XM_006713435.2 | XP_006713498.1 | |||
| XM_011534316.1 | XP_011532618.1 | |||
| AK290554.1 | ||||
| AY137580.1 | AAN11305.1 | |||
| BC007401.2 | AAH07401.1 | |||
| BC018642.2 | AAH18642.1 | |||
| EU812118.1 | ||||
| FJ866789.1 | ||||
| M81933.1 | AAA58415.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001056 | Chr.3:47870852 - 48182359 on Build GRCh38 | Gain |
|
| nsv834682 | Chr.3:48096647 - 48269632 on Build GRCh38 | Gain |
|
| nsv1012024 | Chr.3:48077739 - 48201180 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map