Assay Details
Target Gene Details
Entrez Gene ID: | 5526 |
Gene Name: | protein phosphatase 2 regulatory subunit B'beta |
Gene Aliases: |
B56B, PR61B |
Location: |
Chr.11:64917553-64934478 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R5B | NM_006244.3 | 14 | 2542 | NP_006235.1 |
| XM_011545132.2 | 15 | 1975 | XP_011543434.1 | |
| XM_011545133.1 | 16 | 2095 | XP_011543435.1 | |
| XM_011545134.2 | 14 | 3381 | XP_011543436.1 | |
| AK092730.1 | 9 | 1614 | ||
| BC045619.1 | 14 | 2536 | AAH45619.1 | |
| BM665417.1 | 1 | 225 | ||
| L42374.1 | 14 | 2245 | AAC37602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555208 | Chr.11:64877647 - 65003707 on Build GRCh38 | Loss |
|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map