Assay Details
Target Gene Details
Entrez Gene ID: | 79570 |
Gene Name: | Na+/K+ transporting ATPase interacting 1 |
Gene Aliases: |
FAM77C |
Location: |
Chr.1:31179270-31239887 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKAIN1 | NM_024522.2 | NP_078798.2 | ||
| XM_017002320.1 | 6 | 2477 | XP_016857809.1 | |
| AK022712.1 | BAB14196.1 | |||
| AL832547.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508711 | Chr.1:31085407 - 31192548 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map