Assay Details
Target Gene Details
Entrez Gene ID: | 5739 |
Gene Name: | prostaglandin I2 (prostacyclin) receptor (IP) |
Gene Aliases: |
IP, PRIPR |
Location: |
Chr.19:46610040-46625118 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGIR | NM_000960.3 | NP_000951.1 | ||
| XM_005259093.2 | XP_005259150.1 | |||
| XM_005259095.3 | XP_005259152.1 | |||
| AK315278.1 | ||||
| AY242134.1 | AAO92301.1 | |||
| BC075814.1 | AAH75814.1 | |||
| BC110342.1 | AAI10343.1 | |||
| D25418.1 | BAA05008.1 | |||
| D29634.1 | BAA06110.1 | |||
| DA652875.1 | ||||
| L29016.1 | AAA36448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833849 | Chr.19:46558831 - 46703060 on Build GRCh38 | Loss |
|
| nsv953589 | Chr.19:46601544 - 46636643 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map