Assay Details
Target Gene Details
Entrez Gene ID: | 9166 |
Gene Name: | estrogen receptor binding site associated, antigen, 9 |
Gene Aliases: |
EB9, PDAF |
Location: |
Chr.8:109539700-109565996 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBAG9 | NM_004215.4 | 1 | 187 | NP_004206.1 |
| NM_198120.2 | NP_936056.1 | |||
| XM_017013960.1 | 1 | 109 | XP_016869449.1 | |
| AB007619.1 | 1 | 68 | BAA22572.1 | |
| AF006265.1 | AAB61617.1 | |||
| AK290651.1 | ||||
| AK308950.1 | 1 | 78 | ||
| AL515533.3 | 1 | 78 | ||
| AY515724.1 | AAR97974.1 | |||
| BC005249.1 | 1 | 98 | AAH05249.1 | |
| BC017729.1 | AAH17729.1 | |||
| BC022506.1 | 1 | 85 | AAH22506.1 | |
| BG427211.1 | 1 | 146 | ||
| BG707115.1 | ||||
| BI194881.1 | 1 | 188 | ||
| CD108096.1 | 1 | 131 |
More Information
Set Membership: |
|

Genomic Map