Assay Details
Target Gene Details
Entrez Gene ID: | 102 |
Gene Name: | ADAM metallopeptidase domain 10 |
Gene Aliases: |
AD10, AD18, CD156c, CDw156, HsT18717, MADM, RAK, kuz |
Location: |
Chr.15:58595204-58749978 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM10 | NM_001110.3 | 16 | 2718 | NP_001101.1 |
| NM_001320570.1 | 15 | 2625 | NP_001307499.1 | |
| XM_017021861.1 | 17 | 2773 | XP_016877350.1 | |
| AF009615.1 | 16 | 2743 | AAC51766.1 | |
| AK023460.1 | 4 | 1978 | ||
| AK300472.1 | ||||
| BC126253.1 | 16 | 2548 | ||
| Z48579.1 | 14 | 2103 | CAA88463.1 |
More Information
Set Membership: |
|

Genomic Map