Assay Details
Target Gene Details
Entrez Gene ID: | 675 |
Gene Name: | BRCA2, DNA repair associated |
Gene Aliases: |
BRCC2, BROVCA2, FACD, FAD, FAD1, FANCD, FANCD1, GLM3, PNCA2, XRCC11 |
Location: |
Chr.13:32315480-32399672 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRCA2 | NM_000059.3 | 11 | 5627 | NP_000050.2 |
| DQ897648.1 | 10 | 5400 | ||
| U43746.1 | 11 | 5628 | AAB07223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832576 | Chr.13:32322994 - 32502851 on Build GRCh38 | Gain |
|
| esv3580597 | Chr.13:32133371 - 32386655 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map