Assay Details
Target Gene Details
Entrez Gene ID: | 4324 |
Gene Name: | matrix metallopeptidase 15 |
Gene Aliases: |
MMP-15, MT2-MMP, MT2MMP, MTMMP2, SMCP-2 |
Location: |
Chr.16:58025566-58046901 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP15 | NM_002428.3 | 7 | 2035 | NP_002419.1 |
| BC036495.1 | 7 | 2041 | AAH36495.1 | |
| BC055428.1 | 7 | 2036 | AAH55428.1 | |
| BT006715.1 | 5 | 935 | AAP35361.1 | |
| D85510.1 | 4 | 765 | BAA22225.1 | |
| D86331.1 | 6 | 973 | BAA13071.1 | |
| Z48482.1 | 7 | 1298 | CAA88373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv572760 | Chr.16:58029609 - 58119987 on Build GRCh38 | Loss |
|
| nsv572761 | Chr.16:58035317 - 58046468 on Build GRCh38 | Loss |
|
| esv3638716 | Chr.16:58011317 - 58046443 on Build GRCh38 | Loss |
|
| nsv572762 | Chr.16:58035565 - 58051978 on Build GRCh38 | Loss |
|
| dgv525e214 | Chr.16:58007762 - 58046443 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map