Assay Details
Target Gene Details
Entrez Gene ID: | 80128 |
Gene Name: | tripartite motif containing 46 |
Gene Aliases: |
GENEY, TRIFIC |
Location: |
Chr.1:155173714-155184971 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM46 | NM_001256599.1 | 8 | 1696 | NP_001243528.1 |
| NM_001256600.1 | 9 | 1640 | NP_001243529.1 | |
| NM_001256601.1 | 8 | 1527 | NP_001243530.1 | |
| NM_001282378.1 | 7 | 1424 | NP_001269307.1 | |
| NM_001282379.1 | 8 | 1686 | NP_001269308.1 | |
| NM_025058.4 | 8 | 1686 | NP_079334.3 | |
| NR_046327.1 | 9 | 1724 | ||
| NR_046329.1 | 10 | 1783 | ||
| NR_104150.1 | 9 | 1724 | ||
| XM_011510002.1 | 9 | 1629 | XP_011508304.1 | |
| XM_017002393.1 | 9 | 1800 | XP_016857882.1 | |
| AK026882.1 | 8 | 1597 | BAB15580.1 | |
| AK096158.1 | 8 | 1574 | ||
| AK128139.1 | 10 | 1783 | BAC87293.1 | |
| AK225075.1 | 8 | 1597 | ||
| AK296300.1 | 9 | 1636 | ||
| AK297318.1 | 7 | 1332 | ||
| AK304542.1 | 9 | 1568 | ||
| AK308075.1 | 8 | 1491 | ||
| AK309699.1 | 8 | 1527 | ||
| AK310429.1 | 11 | 1952 | ||
| AY251386.1 | 8 | 1506 | AAP51206.1 | |
| BC069416.1 | 9 | 1620 | AAH69416.1 | |
| BC069568.1 | 9 | 1620 | AAH69568.1 | |
| BC126372.1 | 9 | 1641 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527682 | Chr.1:155177242 - 155227671 on Build GRCh38 | Gain |
|
| nsv470741 | Chr.1:155142784 - 155260340 on Build GRCh38 | Loss |
|
| nsv8447 | Chr.1:155172153 - 155184461 on Build GRCh38 | Gain |
|
| nsv1010146 | Chr.1:155146620 - 155231273 on Build GRCh38 | Gain |
|
| nsv547961 | Chr.1:155109822 - 155225189 on Build GRCh38 | Loss |
|
| dgv3n68 | Chr.1:155122502 - 155343618 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map