Assay Details
Target Gene Details
Entrez Gene ID: | 56751 |
Gene Name: | BarH like homeobox 1 |
Gene Aliases: |
- |
Location: |
Chr.9:132582606-132590253 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BARHL1 | NM_020064.3 | 3 | 1700 | NP_064448.1 |
| AF325688.1 | 3 | 1523 | AAG50279.1 | |
| AK054797.1 | 3 | 1700 | BAB70807.1 |
More Information
Set Membership: |
|

Genomic Map