Assay Details
Target Gene Details
Entrez Gene ID: | 392391 |
Gene Name: | olfactory receptor family 5 subfamily C member 1 |
Gene Aliases: |
OR5C2P, OR9-31, OR9-F |
Location: |
Chr.9:122788933-122789895 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR5C1 | NM_001001923.1 | 1 | 283 | NP_001001923.1 |
| BC136700.1 | 1 | 345 | ||
| BC136701.1 | 1 | 345 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518250 | Chr.9:122779920 - 122794439 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map