Assay Details
Target Gene Details
Entrez Gene ID: | 10382 |
Gene Name: | tubulin beta 4A class IVa |
Gene Aliases: |
DYT4, TUBB4, beta-5 |
Location: |
Chr.19:6494319-6502848 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBB4A | NM_001289123.1 | NP_001276052.1 | ||
| NM_001289127.1 | NP_001276056.1 | |||
| NM_001289129.1 | NP_001276058.1 | |||
| NM_001289130.1 | NP_001276059.1 | |||
| NM_001289131.1 | NP_001276060.1 | |||
| NM_006087.3 | NP_006078.2 | |||
| AK075307.1 | ||||
| AK095202.1 | ||||
| AK293503.1 | ||||
| AK294102.1 | ||||
| AK295917.1 | ||||
| BC006570.2 | AAH06570.1 | |||
| BC013683.1 | AAH13683.1 | |||
| DA265791.1 | ||||
| DA302559.1 | ||||
| DC303150.1 | ||||
| DC314277.1 | ||||
| DC353527.1 | ||||
| DC354036.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv962996 | Chr.19:6495818 - 6502811 on Build GRCh38 | Duplication |
|
| nsv833727 | Chr.19:6400101 - 6568241 on Build GRCh38 | Loss |
|
| nsv833728 | Chr.19:6499789 - 6675421 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map