Assay Details
Target Gene Details
Entrez Gene ID: | 4693 |
Gene Name: | NDP, norrin cystine knot growth factor |
Gene Aliases: |
EVR2, FEVR, ND |
Location: |
Chr.X:43948776-43973675 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDP | NM_000266.3 | 3 | 1683 | NP_000257.1 |
| BC029901.1 | 2 | 1120 | AAH29901.1 | |
| BE139596.1 | 1 | 330 | ||
| X65724.1 | 3 | 1520 | CAA46639.1 | |
| X65882.1 | 3 | 1512 | CAA46713.1 |
More Information
Set Membership: |
|

Genomic Map