Assay Details
Target Gene Details
Entrez Gene ID: | 79066 |
Gene Name: | methyltransferase like 16 |
Gene Aliases: |
METT10D |
Location: |
Chr.17:2416049-2511906 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL16 | NM_024086.3 | 9 | 1090 | NP_076991.3 |
| XM_005256801.2 | 8 | 927 | XP_005256858.1 | |
| XM_017025045.1 | 10 | 1133 | XP_016880534.1 | |
| XM_017025046.1 | 7 | 841 | XP_016880535.1 | |
| AK027410.1 | 9 | 1070 | BAB55094.1 | |
| AK294547.1 | 9 | 1070 | ||
| AK296464.1 | 7 | 815 | ||
| AL832612.2 | 9 | 1049 | CAD89999.2 | |
| BC001213.1 | AAH01213.1 | |||
| BC050603.1 | 9 | 1030 | AAH50603.1 | |
| BX647255.1 | 8 | 864 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574224 | Chr.17:2269769 - 2423186 on Build GRCh38 | Gain |
|
| esv3892947 | Chr.17:2414883 - 2552229 on Build GRCh38 | Gain |
|
| nsv833338 | Chr.17:2363607 - 2592205 on Build GRCh38 | Loss |
|
| nsv111844 | Chr.17:2417086 - 2424120 on Build GRCh38 | Deletion |
|
| nsv1063109 | Chr.17:2342302 - 2591288 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map