Assay Details
Target Gene Details
Entrez Gene ID: | 8934 |
Gene Name: | RAB29, member RAS oncogene family |
Gene Aliases: |
RAB7L, RAB7L1 |
Location: |
Chr.1:205767986-205775487 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB29 | NM_001135662.1 | 4 | 567 | NP_001129134.1 |
| NM_001135663.1 | 2 | 782 | NP_001129135.1 | |
| NM_001135664.1 | 3 | 414 | NP_001129136.1 | |
| NM_003929.2 | 4 | 668 | NP_003920.1 | |
| XM_005245569.1 | 4 | 566 | XP_005245626.1 | |
| XM_005245570.1 | 4 | 619 | XP_005245627.1 | |
| XM_005245571.1 | 4 | 544 | XP_005245628.1 | |
| XM_006711605.3 | 3 | 310 | XP_006711668.1 | |
| XM_006711606.2 | 3 | 274 | XP_006711669.1 | |
| XM_017002748.1 | 3 | 569 | XP_016858237.1 | |
| XM_017002749.1 | 3 | 547 | XP_016858238.1 | |
| XM_017002750.1 | 3 | 479 | XP_016858239.1 | |
| AK223514.1 | 3 | 444 | BAD97234.1 | |
| AK303879.1 | 2 | 533 | ||
| AK308359.1 | 3 | 383 | ||
| AK311085.1 | ||||
| AK313000.1 | 4 | 556 | ||
| BC002585.2 | 4 | 501 | AAH02585.1 | |
| BT019540.1 | 3 | 328 | AAV38347.1 | |
| BX647960.1 | 4 | 633 | ||
| CR542010.1 | 3 | 328 | CAG46807.1 | |
| D84488.1 | 3 | 368 | BAA22160.1 | |
| DA284582.1 | ||||
| DA377295.1 | 3 | 369 |
More Information
Set Membership: |
|

Genomic Map