Assay Details
Target Gene Details
Entrez Gene ID: | 347517 |
Gene Name: | RAB41, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.X:70280978-70285088 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB41 | NM_001032726.2 | 1 | 134 | NP_001027898.2 |
| XM_011530948.2 | 1 | 1328 | XP_011529250.1 | |
| XM_017029488.1 | 1 | 107 | XP_016884977.1 | |
| BC117239.1 | 1 | 134 |
More Information
Set Membership: |
|

Genomic Map