Assay Details
Target Gene Details
Entrez Gene ID: | 11016 |
Gene Name: | activating transcription factor 7 |
Gene Aliases: |
ATFA |
Location: |
Chr.12:53507856-53626415 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF7 | NM_001130060.1 | 9 | 933 | NP_001123532.1 |
| NM_006856.2 | 9 | 996 | NP_006847.1 | |
| NR_073163.1 | 9 | 1029 | ||
| XM_005268587.3 | 9 | 948 | XP_005268644.1 | |
| XM_017018722.1 | 9 | 2136 | XP_016874211.1 | |
| AK298853.1 | 9 | 975 | ||
| AK302907.1 | 7 | 1044 | ||
| AK313507.1 | 9 | 942 | ||
| BC136302.1 | 9 | 882 | ||
| DB047160.1 | ||||
| X52943.1 | 9 | 941 | CAA37118.1 | |
| X57197.1 | 8 | 871 | CAA40483.1 |
More Information
Set Membership: |
|

Genomic Map