Assay Details
Target Gene Details
Entrez Gene ID: | 337979 |
Gene Name: | keratin associated protein 22-1 |
Gene Aliases: |
KAP22.1 |
Location: |
Chr.21:30601121-30601267 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP22-1 | NM_181620.1 | NP_853651.1 | ||
| AB096953.1 | 1 | 42 | BAE46368.1 | |
| BC101682.1 | AAI01683.1 | |||
| BC101686.1 | AAI01687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762105 | Chr.21:30547730 - 30624847 on Build GRCh38 | Gain |
|
| nsv1057467 | Chr.21:30318003 - 30993921 on Build GRCh38 | Gain |
|
| nsv587354 | Chr.21:29978351 - 30620545 on Build GRCh38 | Gain |
|

Genomic Map