Assay Details
Target Gene Details
Entrez Gene ID: | 151354 |
Gene Name: | family with sequence similarity 84 member A |
Gene Aliases: |
NSE1, PP11517 |
Location: |
Chr.2:14632686-14640046 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM84A | NM_145175.2 | 2 | 816 | NP_660158.2 |
| AF370404.1 | 2 | 762 | AAQ15240.1 | |
| AJ417080.1 | 2 | 770 | CAD10038.1 | |
| AK092654.1 | BAC03938.1 | |||
| AK314463.1 | 2 | 785 | ||
| BC026346.1 | 1 | 810 | AAH26346.1 | |
| BC052284.1 | 2 | 765 | AAH52284.1 |

Genomic Map