Assay Details
Target Gene Details
Entrez Gene ID: | 54961 |
Gene Name: | slingshot protein phosphatase 3 |
Gene Aliases: |
SSH3L |
Location: |
Chr.11:67303448-67312607 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSH3 | NM_017857.3 | NP_060327.3 | ||
| XM_017017942.1 | XP_016873431.1 | |||
| AB072360.3 | BAB84119.3 | |||
| AB099291.1 | BAC97814.1 | |||
| AK000522.1 | BAA91228.1 | |||
| AK001790.1 | BAA91913.1 | |||
| AK074432.1 | BAB85080.1 | |||
| AK094226.1 | BAC04314.1 | |||
| BC007709.1 | AAH07709.1 | |||
| DB002847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555253 | Chr.11:67265605 - 67397024 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map