Assay Details
Target Gene Details
Entrez Gene ID: | 3755 |
Gene Name: | potassium voltage-gated channel modifier subfamily G member 1 |
Gene Aliases: |
K13, KCNG, KV6.1, kH2 |
Location: |
Chr.20:51003656-51023176 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNG1 | NM_002237.3 | 3 | 1887 | NP_002228.2 |
| XM_006723785.2 | 5 | 2270 | XP_006723848.1 | |
| XM_011528800.1 | 4 | 1945 | XP_011527102.1 | |
| XM_011528801.1 | 5 | 2054 | XP_011527103.1 | |
| XM_011528802.1 | 5 | 2049 | XP_011527104.1 | |
| XM_011528803.2 | 5 | 1976 | XP_011527105.1 | |
| XM_011528804.1 | 4 | 2199 | XP_011527106.1 | |
| XM_011528805.2 | 3 | 4110 | XP_011527107.1 | |
| XM_011528806.1 | 3 | 2091 | XP_011527108.1 | |
| AF033383.1 | AAC05635.1 | |||
| AK290689.1 | 3 | 1888 | ||
| AW162513.1 | 1 | 340 | ||
| BC046629.2 | 3 | 1856 | AAH46629.1 | |
| CN480356.1 | 1 | 351 |
More Information
Set Membership: |
|

Genomic Map