Assay Details
Target Gene Details
Entrez Gene ID: | 261734 |
Gene Name: | nephronophthisis 4 |
Gene Aliases: |
POC10, SLSN4 |
Location: |
Chr.1:5862808-5992473 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPHP4 | NM_001291593.1 | 21 | 3362 | NP_001278522.1 |
| NM_001291594.1 | 20 | 3185 | NP_001278523.1 | |
| NM_015102.4 | 24 | 3672 | NP_055917.1 | |
| NR_111987.1 | 27 | 4219 | ||
| XM_006710563.3 | 24 | 3518 | XP_006710626.1 | |
| XM_011541213.1 | 23 | 3439 | XP_011539515.1 | |
| XM_011541214.1 | 23 | 3400 | XP_011539516.1 | |
| XM_011541215.1 | 22 | 3331 | XP_011539517.1 | |
| XM_011541216.2 | 25 | 4146 | XP_011539518.1 | |
| XM_011541217.2 | 24 | 4010 | XP_011539519.1 | |
| XM_011541218.2 | 25 | 3679 | XP_011539520.1 | |
| XM_017000996.1 | 23 | 3397 | XP_016856485.1 | |
| XM_017000997.1 | 24 | 3941 | XP_016856486.1 | |
| XM_017000998.1 | 23 | 3442 | XP_016856487.1 | |
| XM_017000999.1 | 24 | 3437 | XP_016856488.1 | |
| XM_017001000.1 | 23 | 3270 | XP_016856489.1 | |
| XM_017001001.1 | 18 | 2709 | XP_016856490.1 | |
| XM_017001003.1 | 16 | 2500 | XP_016856492.1 | |
| AB014573.1 | 19 | 2772 | BAA31648.1 | |
| AF537130.1 | 24 | 3670 | AAN06814.1 | |
| AK124074.1 | 20 | 3138 | ||
| AK127333.1 | 1 | 568 | BAC86935.1 | |
| AK314326.1 | 21 | 3336 | ||
| AY118228.1 | 24 | 3607 | AAM78558.1 | |
| BC040520.1 | 27 | 4220 | AAH40520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545173 | Chr.1:5833391 - 6025293 on Build GRCh38 | Gain |
|
| nsv508723 | Chr.1:5777215 - 5971948 on Build GRCh38 | Insertion |
|
| nsv545174 | Chr.1:5844658 - 5971884 on Build GRCh38 | Loss |
|
| nsv831203 | Chr.1:5830030 - 5967902 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map