Assay Details
Target Gene Details
Entrez Gene ID: | 55118 |
Gene Name: | cartilage acidic protein 1 |
Gene Aliases: |
ASPIC, ASPIC1, CEP-68 |
Location: |
Chr.10:97854711-98030828 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRTAC1 | NM_018058.6 | NP_060528.3 | ||
| XM_011539917.1 | XP_011538219.1 | |||
| XM_017016367.1 | XP_016871856.1 | |||
| AJ276171.1 | CAB98267.1 | |||
| AJ279016.1 | CAC08451.1 | |||
| AK001182.1 | BAA91540.1 | |||
| AK057190.1 | ||||
| AK292084.1 | ||||
| BC034245.1 | AAH34245.1 |
Target Gene Details
Entrez Gene ID: | 401647 |
Gene Name: | golgin A7 family member B |
Gene Aliases: |
C10orf132, C10orf133, bA451M19.3, bA459F3.4 |
Location: |
Chr.10:97850238-97871578 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA7B | NM_001010917.2 | 5 | 2745 | NP_001010917.1 |
| AW172573.1 | 1 | 156 | ||
| BM713854.1 | 1 | 167 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047572 | Chr.10:97839861 - 97877821 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map