Assay Details
Target Gene Details
Entrez Gene ID: | 284656 |
Gene Name: | EPH receptor A10 |
Gene Aliases: |
- |
Location: |
Chr.1:37713888-37765152 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA10 | NM_001099439.1 | NP_001092909.1 | ||
| XM_017001080.1 | XP_016856569.1 | |||
| XM_017001081.1 | XP_016856570.1 | |||
| AK131215.1 | BAD18404.1 | |||
| AK309089.1 | ||||
| AY726560.1 | ||||
| BC041360.1 | AAH41360.1 | |||
| BC112933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525226 | Chr.1:37723169 - 37762544 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map