Assay Details
Target Gene Details
Entrez Gene ID: | 54463 |
Gene Name: | family with sequence similarity 134 member B |
Gene Aliases: |
JK-1, JK1 |
Location: |
Chr.5:16473038-16617058 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM134B | NM_001034850.2 | 9 | 2260 | NP_001030022.1 |
| NM_019000.4 | 7 | 2136 | NP_061873.2 | |
| XM_011514053.2 | 10 | 2319 | XP_011512355.1 | |
| XM_011514054.2 | 8 | 2270 | XP_011512356.1 | |
| XM_011514055.2 | 7 | 2060 | XP_011512357.1 | |
| AK000159.1 | 1 | 1179 | BAA90982.1 | |
| AK024920.1 | 7 | 1966 | BAB15034.1 | |
| AL832438.1 | 9 | 1936 | CAH10610.1 | |
| BC030517.1 | AAH30517.1 |
More Information
Set Membership: |
|

Genomic Map