Assay Details
Target Gene Details
Entrez Gene ID: | 23385 |
Gene Name: | nicastrin |
Gene Aliases: |
ATAG1874 |
Location: |
Chr.1:160343056-160358952 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCSTN | NM_001290184.1 | 12 | 1490 | NP_001277113.1 |
| NM_001290186.1 | 8 | 937 | NP_001277115.1 | |
| NM_015331.2 | 11 | 1351 | NP_056146.1 | |
| XM_011509363.2 | 11 | 1568 | XP_011507665.1 | |
| AF240468.1 | 11 | 1369 | AAG11412.1 | |
| AK296153.1 | 6 | 636 | ||
| AK299142.1 | 8 | 824 | ||
| AK314764.1 | 11 | 1258 | ||
| AY359120.1 | 11 | 1228 | AAQ89478.1 | |
| BC047621.1 | 12 | 1447 | AAH47621.1 | |
| D87442.1 | 11 | 1225 | BAA13383.1 |
More Information
Set Membership: |
|

Genomic Map