Assay Details
Target Gene Details
Entrez Gene ID: | 25821 |
Gene Name: | mitochondrial tRNA translation optimization 1 |
Gene Aliases: |
CGI-02, COXPD10 |
Location: |
Chr.6:73461727-73501456 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTO1 | NM_001123226.1 | 1 | 262 | NP_001116698.1 |
| NM_012123.3 | 1 | 262 | NP_036255.2 | |
| NM_133645.2 | 1 | 262 | NP_598400.1 | |
| XM_006715444.2 | 1 | 57 | XP_006715507.1 | |
| XM_006715445.2 | XP_006715508.1 | |||
| XM_006715446.3 | 1 | 262 | XP_006715509.1 | |
| XM_011535723.1 | 1 | 253 | XP_011534025.1 | |
| XM_011535724.1 | XP_011534026.1 | |||
| XM_011535725.2 | XP_011534027.1 | |||
| XM_017010697.1 | 1 | 57 | XP_016866186.1 | |
| XM_017010698.1 | 1 | 253 | XP_016866187.1 | |
| XM_017010699.1 | XP_016866188.1 | |||
| XM_017010700.1 | 1 | 246 | XP_016866189.1 | |
| AF132937.1 | 1 | 185 | AAD27712.1 | |
| AF319422.2 | 1 | 228 | AAG42814.3 | |
| AF469110.1 | 1 | 228 | AAL82394.1 | |
| AF469111.1 | 1 | 228 | AAL82395.1 | |
| AJ420530.1 | 1 | 209 | ||
| AK074625.1 | 1 | 241 | ||
| AK225828.1 | 1 | 246 | ||
| AL833823.1 | 1 | 106 | CAD38685.1 | |
| AY078985.1 | 1 | 228 | AAL85490.1 | |
| AY078986.1 | 1 | 228 | AAL85491.1 | |
| BC005808.2 | AAH05808.2 | |||
| BC011051.1 | 1 | 203 | ||
| DN993156.1 | 1 | 423 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576177 | Chr.6:73460675 - 73492915 on Build GRCh38 | Gain |
|
| nsv830693 | Chr.6:73339619 - 73506357 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map