Assay Details
Target Gene Details
Entrez Gene ID: | 94233 |
Gene Name: | opsin 4 |
Gene Aliases: |
MOP |
Location: |
Chr.10:86653495-86666460 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPN4 | NM_001030015.2 | NP_001025186.1 | ||
| NM_033282.3 | NP_150598.1 | |||
| XM_017016955.1 | 10 | 2716 | XP_016872444.1 | |
| XM_017016956.1 | 9 | 2683 | XP_016872445.1 | |
| XM_017016957.1 | XP_016872446.1 | |||
| AK310520.1 | 4 | 572 | ||
| BC113558.1 | ||||
| BC143688.1 | ||||
| KT381095.1 | 3 | 380 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1320n54 | Chr.10:86653016 - 86668546 on Build GRCh38 | Gain |
|
| nsv1049754 | Chr.10:86604960 - 86699322 on Build GRCh38 | Gain |
|
| nsv524269 | Chr.10:86654455 - 86680809 on Build GRCh38 | Loss |
|
| dgv116n27 | Chr.10:86653555 - 86682683 on Build GRCh38 | Loss |
|
| dgv948n100 | Chr.10:86646619 - 86714855 on Build GRCh38 | Gain |
|
| nsv551783 | Chr.10:86630480 - 86680809 on Build GRCh38 | Loss |
|
| nsv470968 | Chr.10:86647213 - 86709830 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map