Assay Details
Target Gene Details
Entrez Gene ID: | 1203 |
Gene Name: | ceroid-lipofuscinosis, neuronal 5 |
Gene Aliases: |
NCL |
Location: |
Chr.13:76991924-77002517 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLN5 | NM_006493.2 | 4 | 1487 | NP_006484.1 |
| XM_011534917.2 | 5 | 1517 | XP_011533219.1 | |
| AF068227.1 | 4 | 2751 | AAC27614.1 | |
| AK075109.1 | 4 | 1327 | ||
| AK304001.1 | 5 | 1242 |
Target Gene Details
Entrez Gene ID: | 26224 |
Gene Name: | F-box and leucine rich repeat protein 3 |
Gene Aliases: |
FBL3, FBL3A, FBXL3A |
Location: |
Chr.13:76992597-77027196 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL3 | XM_017020538.1 | XP_016876027.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832653 | Chr.13:76948899 - 77115221 on Build GRCh38 | Loss |
|
| nsv528367 | Chr.13:77000848 - 77001261 on Build GRCh38 | Loss |
|
| nsv832652 | Chr.13:76855107 - 77041507 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map