Assay Details
Target Gene Details
Entrez Gene ID: | 2521 |
Gene Name: | FUS RNA binding protein |
Gene Aliases: |
ALS6, ETM4, FUS1, HNRNPP2, POMP75, TLS |
Location: |
Chr.16:31179444-31194871 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FUS | NM_001170634.1 | NP_001164105.1 | ||
| NM_001170937.1 | 4 | 334 | NP_001164408.1 | |
| NM_004960.3 | 4 | 334 | NP_004951.1 | |
| NR_028388.2 | 4 | 334 | ||
| XM_005255233.4 | XP_005255290.1 | |||
| XM_011545781.1 | 4 | 306 | XP_011544083.1 | |
| AB208902.1 | 4 | 294 | BAD92139.1 | |
| AK098774.1 | ||||
| AK130774.1 | 4 | 306 | ||
| AK290936.1 | 4 | 305 | ||
| AK299129.1 | 4 | 303 | ||
| BC000402.2 | 4 | 297 | AAH00402.1 | |
| BC002459.1 | 4 | 269 | AAH02459.1 | |
| BC026062.2 | 4 | 287 | AAH26062.1 | |
| BT007131.1 | 3 | 229 | AAP35795.1 | |
| BX423519.2 | 4 | 313 | ||
| CR456747.1 | 3 | 229 | CAG33028.1 | |
| DA541747.1 | 4 | 305 | ||
| DC361263.1 | 4 | 334 | ||
| S62140.1 | 4 | 307 | AAB27102.1 | |
| U36561.1 | 3 | 226 | AAA79948.1 | |
| X71428.1 | CAA50559.1 |
More Information
Set Membership: |
|

Genomic Map