Assay Details
Target Gene Details
Entrez Gene ID: | 9074 |
Gene Name: | claudin 6 |
Gene Aliases: |
- |
Location: |
Chr.16:3014712-3018187 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLDN6 | NM_021195.4 | NP_067018.2 | ||
| AK075329.1 | ||||
| BC008934.2 | AAH08934.1 | |||
| BX444798.2 | ||||
| CN420342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509588 | Chr.16:2919880 - 3043776 on Build GRCh38 | Insertion |
|
| nsv952906 | Chr.16:2868600 - 3028099 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map