Assay Details
Target Gene Details
Entrez Gene ID: | 6745 |
Gene Name: | signal sequence receptor subunit 1 |
Gene Aliases: |
TRAPA |
Location: |
Chr.6:7281143-7313314 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSR1 | NM_001292008.1 | 8 | 1095 | NP_001278937.1 |
| NM_003144.4 | 8 | 1299 | NP_003135.2 | |
| NR_120448.1 | 7 | 1220 | ||
| AF156965.1 | 8 | 1216 | AAD48778.1 | |
| AK075562.1 | 7 | 1083 | BAC11701.1 | |
| AK222762.1 | 8 | 1167 | BAD96482.1 | |
| AK222809.1 | BAD96529.1 | |||
| AK291550.1 | 8 | 1167 | ||
| AL550910.3 | ||||
| BC007710.2 | 8 | 1192 | AAH07710.1 | |
| CR936719.1 | 7 | 1158 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470797 | Chr.6:7288064 - 7546171 on Build GRCh38 | Gain |
|
| nsv1029748 | Chr.6:7271371 - 7310026 on Build GRCh38 | Gain |
|
| nsv462623 | Chr.6:7278096 - 7546171 on Build GRCh38 | Gain |
|
| esv3890787 | Chr.6:6996928 - 7981438 on Build GRCh38 | Loss |
|
| nsv981350 | Chr.6:7277821 - 7548705 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146482489] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map