Assay Details
Target Gene Details
Entrez Gene ID: | 136 |
Gene Name: | adenosine A2b receptor |
Gene Aliases: |
ADORA2 |
Location: |
Chr.17:15927781-15975896 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADORA2B | NM_000676.2 | 2 | 1028 | NP_000667.1 |
| XM_011523659.2 | 3 | 859 | XP_011521961.1 | |
| XM_017024197.1 | 3 | 805 | XP_016879686.1 | |
| AK172851.1 | 1 | 1179 | ||
| AY136748.1 | 2 | 696 | AAN01274.1 | |
| BC025722.1 | 2 | 782 | AAH25722.1 | |
| M97759.1 | 2 | 831 | AAA51598.1 | |
| X68487.1 | 2 | 1028 | CAA48505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3108n100 | Chr.17:15966243 - 16193927 on Build GRCh38 | Gain |
|
| esv3892973 | Chr.17:15947882 - 16030711 on Build GRCh38 | Gain |
|
| esv3572431 | Chr.17:15951820 - 16194089 on Build GRCh38 | Gain |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
| dgv3107n100 | Chr.17:15703953 - 16150198 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map